SY3H3D67F6AD Synthonix Corporation

1-Methyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 154471-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/40884.html

Safety Information

MSDS https://synthonix.com/40884.html
Availability details for
Quantity
  • Availability
  • Add to Cart