SY3H3D67F6AF Synthonix Corporation

2-Methyl-4-(trifluoromethyl)thiazol-5-amine

  • MDL number No MDL found
  • Cas Number : 958452-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40886.html

Safety Information

MSDS https://synthonix.com/40886.html
Availability details for
Quantity
  • Availability
  • Add to Cart