SY3H3D67F6B8 Synthonix Corporation

METHYL 5-NITRO-1H-IMIDAZOLE-4-CARBOXYLATE

  • MDL number No MDL found
  • Cas Number : 20271-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40981.html

Safety Information

MSDS https://synthonix.com/40981.html
Availability details for
Quantity
  • Availability
  • Add to Cart