SY3H3D67F6BA Synthonix Corporation

Methyl thiazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 55842-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40994.html

Safety Information

MSDS https://synthonix.com/40994.html
Availability details for
Quantity
  • Availability
  • Add to Cart