SY3H3D67F6BE Synthonix Corporation

Methyl oxazole-2-carboxylate

  • Cas Number : 31698-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40998.html

Safety Information

MSDS https://synthonix.com/40998.html
Availability details for
Quantity
  • Availability
  • Add to Cart