SY3H3D67F6C0 Synthonix Corporation

5-Methoxyoxazole-2-carboxylic acid

  • Cas Number : 439109-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41003.html

Safety Information

MSDS https://synthonix.com/41003.html
Availability details for
Quantity
  • Availability
  • Add to Cart