SY3H3D67FAA8 Synthonix Corporation

3-Methylpent-1-yn-3-amine hydrochloride

  • Cas Number : 108575-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43435.html

Safety Information

MSDS https://synthonix.com/43435.html
Availability details for
Quantity
  • Availability
  • Add to Cart