SY3H3D67FAA9 Synthonix Corporation

4-Methyl-5-oxohex-2-enedioic acid

  • MDL number No MDL found
  • Cas Number : 412324-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46274.html

Safety Information

MSDS https://synthonix.com/46274.html
Availability details for
Quantity
  • Availability
  • Add to Cart