SY3H3D67FAAC Synthonix Corporation

4-Methoxypyridazine

  • MDL number No MDL found
  • Cas Number : 20733-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/41023.html

Safety Information

MSDS https://synthonix.com/41023.html
Availability details for
Quantity
  • Availability
  • Add to Cart