SY3H3D67FAB4 Synthonix Corporation

4-Methylpyrazole-3-carboxylic Acid

  • MDL number No MDL found
  • Cas Number : 82231-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41036.html

Safety Information

MSDS https://synthonix.com/41036.html
Availability details for
Quantity
  • Availability
  • Add to Cart