SY3H3D67FAB9 Synthonix Corporation

2-(Methylsulfinyl)pyrimidine

  • MDL number No MDL found
  • Cas Number : 14080-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41047.html

Safety Information

MSDS https://synthonix.com/41047.html
Availability details for
Quantity
  • Availability
  • Add to Cart