SY3H3D67FABD Synthonix Corporation

1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine

  • MDL number No MDL found
  • Cas Number : 1006436-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41085.html

Safety Information

MSDS https://synthonix.com/41085.html
Availability details for
Quantity
  • Availability
  • Add to Cart