SY3H3D67FABE Synthonix Corporation

2-Methyl-5-trifluoromethyl-2H-pyrazol-3-ylamine

  • MDL number No MDL found
  • Cas Number : 149978-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/41086.html

Safety Information

MSDS https://synthonix.com/41086.html
Availability details for
Quantity
  • Availability
  • Add to Cart