SY3H3D67FAC2 Synthonix Corporation

3-Methyldihydrofuran-2,5-dione

  • MDL number MFCD00005527
  • Cas Number : 4100-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41122.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41122.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart