SY3H3D67FAC9 Synthonix Corporation

1-Methyl-1H-imidazole-4-carboxamide

  • Cas Number : 129993-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41145.html

Safety Information

MSDS https://synthonix.com/41145.html
Availability details for
Quantity
  • Availability
  • Add to Cart