SY3H3D67FACF Synthonix Corporation

Methyl 4,4,4-trichlorobutanoate

  • Cas Number : 19376-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41178.html

Safety Information

MSDS https://synthonix.com/41178.html
Availability details for
Quantity
  • Availability
  • Add to Cart