SY3H3D67FAD7 Synthonix Corporation

2-(Methylthio)pyrimidine-4,6-diamine

  • Cas Number : 1005-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41215.html

Safety Information

MSDS https://synthonix.com/41215.html
Availability details for
Quantity
  • Availability
  • Add to Cart