SY3H3D67FAD8 Synthonix Corporation

4-Methoxybut-3-en-2-one

  • MDL number MFCD00008778
  • Cas Number : 4652-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41222.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41222.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart