SY3H3D67FADF Synthonix Corporation

3-Methylpyrrolidin-2-one

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41257.html

Safety Information

MSDS https://synthonix.com/41257.html
Availability details for
Quantity
  • Availability
  • Add to Cart