SY3H3D67FAE5 Synthonix Corporation

(R)-Methyl 4-chloro-3-hydroxybutanoate

  • MDL number No MDL found
  • Cas Number : 88496-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41276.html

Safety Information

MSDS https://synthonix.com/41276.html
Availability details for
Quantity
  • Availability
  • Add to Cart