SY3H3D67FAE6 Synthonix Corporation

6-Methylpyridazin-3(2H)-one Hydrobromide

  • MDL number No MDL found
  • Cas Number : 94248-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41309.html

Safety Information

MSDS https://synthonix.com/41309.html
Availability details for
Quantity
  • Availability
  • Add to Cart