SY3H3D67FAE7 Synthonix Corporation

(2-Methyloxazol-4-yl)methanamine hydrobromide

  • MDL number No MDL found
  • Cas Number : 2061979-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41310.html

Safety Information

MSDS https://synthonix.com/41310.html
Availability details for
Quantity
  • Availability
  • Add to Cart