SY3H3D67FAEE Synthonix Corporation

Methyl 2-aminobutanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 7682-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41330.html

Safety Information

MSDS https://synthonix.com/41330.html
Availability details for
Quantity
  • Availability
  • Add to Cart