SY3H3D67FAF5 Synthonix Corporation

Methyl 1H-imidazole-5-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 127607-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41348.html

Safety Information

MSDS https://synthonix.com/41348.html
Availability details for
Quantity
  • Availability
  • Add to Cart