SY3H3D67FB01 Synthonix Corporation

Methyl 6-oxopiperazine-2-carboxylate

  • MDL number MFCD13175115
  • Cas Number : 234109-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41440.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41440.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart