SY3H3D67FB02 Synthonix Corporation

(S)-Methyl 5-oxopiperazine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1191885-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/41441.html

Safety Information

MSDS https://synthonix.com/41441.html
Availability details for
Quantity
  • Availability
  • Add to Cart