SY3H3D67FB07 Synthonix Corporation

Methyl 2,2-dimethyl-3-oxopropanoate

  • MDL number No MDL found
  • Cas Number : 13865-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41471.html

Safety Information

MSDS https://synthonix.com/41471.html
Availability details for
Quantity
  • Availability
  • Add to Cart