SY3H3D67FB09 Synthonix Corporation

(R)-4-Methoxy-2-methyl-4-oxobutanoic acid

  • MDL number No MDL found
  • Cas Number : 81025-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41477.html

Safety Information

MSDS https://synthonix.com/41477.html
Availability details for
Quantity
  • Availability
  • Add to Cart