SY3H3D67FB0C Synthonix Corporation

(1-(2-Methoxyethyl)-1H-pyrazol-4-yl)boronic acid

  • Cas Number : 847818-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41488.html

Safety Information

MSDS https://synthonix.com/41488.html
Availability details for
Quantity
  • Availability
  • Add to Cart