SY3H3D67FB0D Synthonix Corporation

4-Methyl-3-propoxy-1H-1,2,4-triazol-5(4H)-one

  • MDL number No MDL found
  • Cas Number : 145027-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41494.html

Safety Information

MSDS https://synthonix.com/41494.html
Availability details for
Quantity
  • Availability
  • Add to Cart