SY3H3D67FB0F Synthonix Corporation

Methyl 3-(methylamino)but-2-enoate

  • MDL number No MDL found
  • Cas Number : 13412-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41506.html

Safety Information

MSDS https://synthonix.com/41506.html
Availability details for
Quantity
  • Availability
  • Add to Cart