SY3H3D67FB11 Synthonix Corporation

(R)-Methyl 2-acetamidopropanoate

  • MDL number No MDL found
  • Cas Number : 19914-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41515.html

Safety Information

MSDS https://synthonix.com/41515.html
Availability details for
Quantity
  • Availability
  • Add to Cart