SY3H3D67FB15 Synthonix Corporation

2-Methoxyhexahydropyrimidine-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 1956319-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41538.html

Safety Information

MSDS https://synthonix.com/41538.html
Availability details for
Quantity
  • Availability
  • Add to Cart