SY3H3D67FB17 Synthonix Corporation

(S)-3-Methylvaleric Acid

  • MDL number No MDL found
  • Cas Number : 1730-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41552.html

Safety Information

MSDS https://synthonix.com/41552.html
Availability details for
Quantity
  • Availability
  • Add to Cart