SY3H3D67FB21 Synthonix Corporation

(trans-4-Methylpyrrolidin-3-yl)methanol

  • MDL number No MDL found
  • Cas Number : 945723-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41583.html

Safety Information

MSDS https://synthonix.com/41583.html
Availability details for
Quantity
  • Availability
  • Add to Cart