SY3H3D67FB22 Synthonix Corporation

(2R)-2-(Methoxymethyl)morpholine

  • Cas Number : 157791-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41585.html

Safety Information

MSDS https://synthonix.com/41585.html
Availability details for
Quantity
  • Availability
  • Add to Cart