SY3H3D67FB26 Synthonix Corporation

Methyl 4,6-dichloropyrimidine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 811450-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42102.html

Safety Information

MSDS https://synthonix.com/42102.html
Availability details for
Quantity
  • Availability
  • Add to Cart