SY3H3D67FB28 Synthonix Corporation

2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 117724-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42257.html

Safety Information

MSDS https://synthonix.com/42257.html
Availability details for
Quantity
  • Availability
  • Add to Cart