SY3H3D67FB2B Synthonix Corporation

Methyl 4,5-dichlorothiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 89281-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42320.html

Safety Information

MSDS https://synthonix.com/42320.html
Availability details for
Quantity
  • Availability
  • Add to Cart