SY3H3D67FB2C Synthonix Corporation

Methyl 4,5-dibromo-2-furoate

  • MDL number No MDL found
  • Cas Number : 54113-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42328.html

Safety Information

MSDS https://synthonix.com/42328.html
Availability details for
Quantity
  • Availability
  • Add to Cart