SY3H3D67FB2D Synthonix Corporation

Methyl 4,5-dichloro-3-hydroxythiophene-2-carboxylate

  • Cas Number : 96232-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42330.html

Safety Information

MSDS https://synthonix.com/42330.html
Availability details for
Quantity
  • Availability
  • Add to Cart