SY3H3D67FB2E Synthonix Corporation

Methyl 5-chloro-3-(chlorosulfonyl)thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 126910-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42335.html

Safety Information

MSDS https://synthonix.com/42335.html
Availability details for
Quantity
  • Availability
  • Add to Cart