SY3H3D67FB30 Synthonix Corporation

2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one

  • MDL number No MDL found
  • Cas Number : 175354-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42422.html

Safety Information

MSDS https://synthonix.com/42422.html
Availability details for
Quantity
  • Availability
  • Add to Cart