SY3H3D67FB32 Synthonix Corporation

Methyl 3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 61859-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42424.html

Safety Information

MSDS https://synthonix.com/42424.html
Availability details for
Quantity
  • Availability
  • Add to Cart