SY3H3D67FB36 Synthonix Corporation

2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol

  • Cas Number : 2836-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42465.html

Safety Information

MSDS https://synthonix.com/42465.html
Availability details for
Quantity
  • Availability
  • Add to Cart