SY3H3D67FB37 Synthonix Corporation

6-Methyl-2-(trifluoromethyl)pyrimidin-4(3H)-one

  • MDL number No MDL found
  • Cas Number : 2557-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42466.html

Safety Information

MSDS https://synthonix.com/42466.html
Availability details for
Quantity
  • Availability
  • Add to Cart