SY3H3D67FB38 Synthonix Corporation

5-Methylpyrazine-2-carbonitrile

  • MDL number No MDL found
  • Cas Number : 98006-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42468.html

Safety Information

MSDS https://synthonix.com/42468.html
Availability details for
Quantity
  • Availability
  • Add to Cart