SY3H3D67FB41 Synthonix Corporation

Methyl 2-bromothiophene-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 76360-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42697.html

Safety Information

MSDS https://synthonix.com/42697.html
Availability details for
Quantity
  • Availability
  • Add to Cart