SY3H3D67FB46 Synthonix Corporation

Methyl 5-iodothiophene-2-carboxylate

  • Cas Number : 88105-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42708.html

Safety Information

MSDS https://synthonix.com/42708.html
Availability details for
Quantity
  • Availability
  • Add to Cart