SY3H3D67FB47 Synthonix Corporation

Methyl 4-bromofuran-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 58235-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42715.html

Safety Information

MSDS https://synthonix.com/42715.html
Availability details for
Quantity
  • Availability
  • Add to Cart